Learn More
Quinaldine, 95+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 132170010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quinaldine | |
91-63-4 | |
248.0°C | |
Authentic | |
Glass bottle | |
1.6070 to 1.6120 | |
1L | |
1.058 | |
quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
2-methylquinoline | |
7060 | |
95+% |
97% | |
1.0580g/mL | |
79°C | |
95% min. (GC) | |
C10H9N | |
MFCD00006756 | |
20, 387 | |
15, 8163 | |
Solubility in water: 2.5g/L (20°C). Other solubilities: soluble in chloroform,ether and alcohol | |
CC1=CC=C2C=CC=CC2=N1 | |
143.19 | |
143.19 |
Chemical Identifiers
91-63-4 | |
143.19 | |
SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
7060 | |
CC1=CC=C2C=CC=CC2=N1 |
C10H9N | |
MFCD00006756 | |
quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
2-methylquinoline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doct
GHS Signal Word: Warning
EINECSNumber : 202-085-1
RTECSNumber : UZ9625000
TSCA : TSCA
Recommended Storage : Product may discolor to brownish on storage
RUO â Research Use Only