missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinaldic acid, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC171050500

 View more versions of this product

Catalog No. AC171050500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Quinaldic acid
Glass bottle
159.0°C to 161.0°C
93-10-7
C10H7NO2
22, 71
quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
C1=CC=C2C(=C1)C=CC(=N2)C(=O)O
173.171
CHEBI:18386
Needle-Like Crystalline Powder
Authentic
Brown
50g
99%
MFCD00006752
15, 8162
LOAUVZALPPNFOQ-UHFFFAOYSA-N
quinoline-2-carboxylic acid
7124
173.17
99%

Chemical Identifiers

93-10-7
173.171
LOAUVZALPPNFOQ-UHFFFAOYSA-N
7124
quinoline-2-carboxylic acid
C10H7NO2
MFCD00006752
quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
CHEBI:18386
C1=CC=C2C(=C1)C=CC(=N2)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents