missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinaldic acid, 98%

Manufacturer:  Alfa Aesar™ A1019206

 View more versions of this product

Catalog No. AAA1019206



Specifications

Specifications

Quinaldic acid
98%
MFCD00006752
14,8046
LOAUVZALPPNFOQ-UHFFFAOYSA-N
quinoline-2-carboxylic acid
7124
173.17
5g
93-10-7
C10H7NO2
126322
Quinoline-2-carboxylic acid
C1=CC=C2C(=C1)C=CC(=N2)C(=O)O
173.171
CHEBI:18386
156°C to 159°C

Chemical Identifiers

93-10-7
173.171
LOAUVZALPPNFOQ-UHFFFAOYSA-N
7124
quinoline-2-carboxylic acid
C10H7NO2
MFCD00006752
Quinoline-2-carboxylic acid
CHEBI:18386
C1=CC=C2C(=C1)C=CC(=N2)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UZ9100000

TSCA : TSCA

SDS Documents