missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quinaldic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1019222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quinaldic acid | |
156°C to 159°C | |
MFCD00006752 | |
126322 | |
quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid | |
OC(=O)C1=CC=C2C=CC=CC2=N1 | |
173.17 | |
CHEBI:18386 | |
98% |
93-10-7 | |
C10H7NO2 | |
100 g | |
14,8046 | |
LOAUVZALPPNFOQ-UHFFFAOYSA-N | |
quinoline-2-carboxylic acid | |
7124 | |
173.17 |
Chemical Identifiers
93-10-7 | |
173.17 | |
LOAUVZALPPNFOQ-UHFFFAOYSA-N | |
7124 | |
quinoline-2-carboxylic acid |
C10H7NO2 | |
MFCD00006752 | |
quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid | |
CHEBI:18386 | |
OC(=O)C1=CC=C2C=CC=CC2=N1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-218-3
RTECSNumber : UZ9100000
TSCA : Yes
RUO – Research Use Only