Please login to your online account to display your discounted pricing
Quidel™ QuickVue™ H. pylori g II™ Test
Rapid immunoassay for the simple, accurate and qualitative detection of IgG antibodies specific to H. pylori
Manufacturer: quidel  Q0W009
 View more versions of this product
Catalog No. 120116
  • / Pack
Description & Specifications
Specifications
Clia Complexity Waived, non-waived
Detectable Analytes IgG Antibodies specific to H. pylori
Includes Testing cassettes, positive and negative control, transfer pipets and capillary tubes, package insert and procedure card, positive and negative external control
Quantity 10 Tests
Shelf Life 1 year
Storage Requirements Room temperature
Product Type H. pylori gII Test Kit
Accessories
1 year