missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quetiapine Hemifumarate 98.0+%, TCI America™

Manufacturer:  Tci America Q009225G

 View more versions of this product

Catalog No. Q009225G



Specifications

Specifications

Quetiapine Hemifumarate
174°C
111974-72-2
MFCD03423782
ZTHJULTYCAQOIJ-KSBRXOFISA-N
2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethanol;(Z)-but-2-enedioic acid
45358180
Crystal-Powder at 20°C
White-Yellow
25g
C46H54N6O8S2
quetiapine fumarate, 2-methoxyethanol; quetiapine
C1CN(CCN1CCOCCO)C2=NC3=CC=CC=C3SC4=CC=CC=C42.C1CN(CCN1CCOCCO)C2=NC3=CC=CC=C3SC4=CC=CC=C42.C(=CC(=O)O)C(=O)O
883.092
441.55
≥98.0% (HPLC,T)

Chemical Identifiers

111974-72-2
883.092
ZTHJULTYCAQOIJ-KSBRXOFISA-N
45358180
C1CN(CCN1CCOCCO)C2=NC3=CC=CC=C3SC4=CC=CC=C42.C1CN(CCN1CCOCCO)C2=NC3=CC=CC=C3SC4=CC=CC=C42.C(=CC(=O)O)C(=O)O
C46H54N6O8S2
MFCD03423782
quetiapine fumarate, 2-methoxyethanol; quetiapine
2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethanol;(Z)-but-2-enedioic acid
Safety and Handling

Safety and Handling

RTECSNumber : KK3605000

TSCA : No

Documents