Invitrogen™ Qubit™ 1X dsDNA HS Assay Kit DFS Item
The Qubit 1X dsDNA HS (High-Sensitivity) Assay Kit is designed specifically for use with the Qubit Fluorometer.
$149.00 - $449.00
Specifications
Applications Designed specifically for use with the Qubit Fluorometer.
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
Q33230 View Documents Invitrogen™
LSQ33230
100 assays Each for $149.00
Q33231 View Documents Invitrogen™
LSQ33231
500 assays Each for $449.00
Description
Description
The assay is highly selective for double-stranded DNA (dsDNA) over RNA and is designed to be accurate for initial sample concentrations from 10pg/µL to 100ng/µL. The “1X” dsDNA assay kit provides reagent and buffer in a formulation that is stable as a ready-to-use solution for up to one year after preparation. Simply add your sample (any volume between 1 µL and 20 µL is acceptable) and read the concentration using a Qubit 2.0, 3, or 4 fluorometer. Common contaminants such as salts, free nucleotides, solvents, detergents, and protein are well-tolerated in the assay.
Notes:
  1. The Qubit 1X dsDNA HS Assay works on Qubit 2.0, 3, and 4 fluorometers. It will not work on the original Qubit 1.0 Fluorometer.
  2. 500mL thin-walled PCR tubes (Cat. No. Q32856) are required but not included.
Specifications
Specifications
Designed specifically for use with the Qubit Fluorometer.
For Research Use Only. Not for use in diagnostic procedures.