Please login to your online account to display your discounted pricing
Quantum™ Big Screen Microscope
Easy viewing of specimens.
$18.25 - $18.25
Specifications
Includes Assortment of accessories, Glare-reducing shield, Snap-Tite tray to hold components
Required Accessories Two C battery or AC adapter
View More Specs
Includes: 
  • Assortment of accessories
  • Glare-reducing shield
  • Snap-Tite™ tray to hold components
Requires:  Two “C” batteries or AC adapter
Products
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity    
S79033
fisher scientific
LER2901
AC adapter for quantum big screen microscope AC adapter Each for $18.25
Description & Specifications
Specifications
Includes Assortment of accessories, Glare-reducing shield, Snap-Tite tray to hold components
Required Accessories Two C battery or AC adapter
This 7 in. (17.5cm) screen microscope projects a 10X, 20X, or 40X clear image.