Applied Biosystems™ QuantStudio™ 7 Flex Real-Time PCR System, 384-well DFS Item
Advances your research further by enabling a broad range of real-time PCR-based applications through its multiplexing capabilities and interchangeable block formats.
Specifications
Quantity 1 System
Thermal Block Formats 96-well plate, Interchangeable
For Use With (Application) Digital PCR, genotyping and genomic profiling, real time PCR (qPCR), real time PCR-based gene expression profiling
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
4485693 View Documents Applied Biosystems™
LS4485693
Desktop computer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4485698 View Documents Applied Biosystems™
LS4485698
Notebook Computer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
With a simplified workflow, intuitive software, automation capabilities, and touch-screen interface, the QuantStudio™ 7 Flex system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.
Features of the QuantStudio™ 7 Flex Real-Time PCR System include:
  • Increased application versatility through the accommodation of 4 different block types
  • Compatible with over 8 million TaqMan™ assays for gene exp
Specifications
Specifications
1 System
96-well plate, Interchangeable
Digital PCR, genotyping and genomic profiling, real time PCR (qPCR), real time PCR-based gene expression profiling