Applied Biosystems™ QuantStudio™ 5 Real-Time PCR System, 96-well DFS Item
Designed for users who need superior performance, maximum dye versatility, and security options in a real-time PCR system that is affordable and easy to use.
Specifications
For Use With (Application) Real time PCR (qPCR)
View More Specs
Products 6
Catalog Number Mfr. No. Includes Volume (Metric) Thermal Block Sample Price Quantity  
Catalog Number Mfr. No. Includes Volume (Metric) Thermal Block Sample Price Quantity  
A28138 View Documents Applied Biosystems™
LSA28138
No Computer 0.1mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28573 View Documents Applied Biosystems™
LSA28573
Desktop Computer 0.1mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28568 View Documents Applied Biosystems™
LSA28568
Laptop Computer 0.1mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28574 View Documents Applied Biosystems™
LSA28574
Desktop computer 0.2mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28569 View Documents Applied Biosystems™
LSA28569
Laptop Computer 0.2mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28139 View Documents Applied Biosystems™
LSA28139
No Computer 0.2mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The optimized Design and Analysis software is ideal for both first-time and experienced users. When connected to the Thermo Fisher Cloud, the QuantStudio™ 5 system provides access to your data wherever and whenever you want.
Features of the QuantStudio™ 5 system with 96-well block include:
  • Proven OptiFlex™ technology (6 decoupled channels/21 filter combinations with white LED) for trusted results
  • User account
    Order Info
    Room Temperature
Specifications
Specifications
Real time PCR (qPCR)