Applied Biosystems™ QuantStudio™ 3 Real-Time PCR System, 96-well DFS Item
QuantStudio™ 3 Real-Time PCR System is designed for users who need an affordable, easy-to-use real-time PCR system that doesn't compromise performance and quality.
Specifications
For Use With (Application) Real time PCR (qPCR)
View More Specs
Products 5
Catalog Number Mfr. No. Includes Volume (Metric) Thermal Block Sample Price Quantity  
Catalog Number Mfr. No. Includes Volume (Metric) Thermal Block Sample Price Quantity  
A28137 View Documents Applied Biosystems™
LSA28137
Computer not included 0.2mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28136 View Documents Applied Biosystems™
LSA28136
Computer not included 0.1mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28566 View Documents Applied Biosystems™
LSA28566
Training included 0.1mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28572 View Documents Applied Biosystems™
LSA28572
Desktop computer 0.2mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A28567 View Documents Applied Biosystems™
LSA28567
Laptop Computer 0.2mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The simplified Design and Analysis software is ideal for both first-time and experienced users. When connected to the Thermo Fisher Cloud, the QuantStudio™ 3 system provides access to your data wherever and whenever you want.
Features of the QuantStudio™ 3 system include:
  • Simplified instrument software with interactive touchscreen for greater ease of use
  • Proven OptiFlex™ technology (4 coupled channels with white LED) for trusted resu
    Order Info
    Room Temperature
Specifications
Specifications
Real time PCR (qPCR)