missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR System, OpenArray™ Block, AccuFill™ System DFS Item

QuantStudio™ 12K Flex system is a highly flexible, comprehensive RT-PCR platform that leverages the best of Applied Biosystems™ real-time technology in a single instrument.

Specifications

orderInfo Room Temperature
Quantity 1 System
Item Description Power limit: 60Hz
Height (Metric) 74cm
Depth (Metric) 67cm
View More Specs

Includes: With OpenArray™Block; AccuFill™System; Desktop Computer

Products 4
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
4471086 View Documents Applied Biosystems™
LS4471086
Laptop N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4471090 View Documents Applied Biosystems™
LS4471090
Without Power Cord N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4472380 View Documents Applied Biosystems™
LS4472380
Desktop Computer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4472379 View Documents Applied Biosystems™
LS4472379
Laptop Computer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

The QuantStudio™ 12K Flex Real-Time PCR System includes easy-to-change blocks that can accommodate OpenArray™ plates, TaqMan™ array cards, 384-well, 96-well, and FAST 96-well plates allowing you to select the format that is right for you

Thermo Fisher Cloud:
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast
Specifications

Specifications

Room Temperature
Power limit: 60Hz
67cm
High throughput-compatible, multiplexing
21 Color options, 6 emission filters (500 to 720nm), 6 excitation filters (450 to 670nm)
Down to 1 copy
69kg
1 System
74cm
Up to 9 logs of linear dynamic range
100 to 240VAC
1μL (384-well array cards)
152 lb.
Digital PCR, genotyping and genomic profiling, real time PCR (qPCR), real time PCR-based gene expression profiling