missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ QuantaBlu™ Fluorogenic Peroxidase Substrate

Conduct stopped, non-stopped and kinetic assays using this soluble fluorogenic (chemifluorescent) peroxidase substrate for ELISA.

$284.20 - $379.41

Specifications

Kit Contents

QuantaBlu Peroxide Solution, 30mL

,

QuantaBlu Peroxide Solution, 30mL

Sufficient For 280mL of 1X substrate buffer
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15162 Thermo Scientific™
15162*
QuantaBlu NS/K Substrate (for non-stopped and kinetic assay) Each for $284.20
PI15169 Thermo Scientific™
PI15169
QuantaBlu Fluorogenic Peroxidase Substrate Each for $379.41
Specifications

Description

Thermo Scientific™ QuantaBlu Fluorogenic Peroxidase Substrate is a soluble fluorogenic (chemifluorescent) substrate for detection of peroxidase activity in ELISA methods.


Additional Information and Online Ordering »

In the presence of active peroxidase the working solution of QuantaBlu Substrate produces a blue fluorescent product that can be quantitated by fluorometry in microplates or cuvettes. Fluorometric-based detection has a large dynamic range, which overcomes the limitations of using spectrophotometry to measure colorimetric substrates. The blue fluorescent product is not sensitive to light and does not photobleach. QuantaBlu Fluorogenic Peroxidase Substrate is stable, highly sensitive, produces high signal-to-noise ratios and allows for a broad dynamic detection range of peroxidase activity. This substrate is also flexible and can be used for stopped, nonstopped and kinetic assays. Incubation for stopped and nonstopped assays can be performed between 5 and 90 minutes at either room temperature (RT) or 37°C. The QuantaBlu NS/K Substrate formulation can be integrated into robotic-based assays and is designed for assays that do not need to be stopped.

Highlights:

  • More sensitive than TMB, OPD or ABTS substrates
  • Flexible stopped, nonstopped or kinetic assays possible
  • Large dynamic range (4 log peroxidase concentration range)
  • Excellent stability; working solution is stable for 24 hours
  • Large stokes shift; excitation/emission maxima of 325nm/420nm (range of 315-345/370-460)
  • Does not photobleach
Description & Specifications

Specifications

QuantaBlu Peroxide Solution, 30mL

,

QuantaBlu Peroxide Solution, 30mL

280mL of 1X substrate buffer