Please login to your online account to display your discounted pricing

Quality Control Standard 27, Ricca Chemical


Quantity 500mL
Packaging Amber poly

Manufacturer: ricca chemical company  RISQCSTD27500B

 View more versions of this product

Catalog No. SQCSTD27500

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Clear light colored
Physical Form Liquid
Quantity 500mL
Chemical Name or Material Quality Control Standard 27
Shelf Life 12 months
CAS Max % 0.05
CAS 7789-02-8
CAS Min % 0.03
CAS Max % 0.03
CAS 16919-19-0
CAS Min % 0.02
CAS Max % 0.02
CAS 10043-35-3
CAS Min % 0.02
CAS Max % 0.02
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
Packaging Amber poly
CAS Min % 3.14
CAS Max % 3.27
CAS 7784-27-2
CAS Min % 0.05

Specifications

Chemical Name or Material Quality Control Standard 27
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 3.14
CAS Max % 3.27
CAS 7784-27-2
CAS Min % 0.05
CAS Max % 0.05
CAS 7789-02-8
CAS Min % 0.03
CAS Max % 0.03
CAS 16919-19-0
CAS Min % 0.02
CAS Max % 0.02
CAS 10043-35-3
CAS Min % 0.02
CAS Max % 0.02
Physical Form Liquid
Accessories

12 months