missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quality Control Standard 27, Ricca Chemical

Manufacturer:  Ricca Chemical Company RISQCSTD27100B

 View more versions of this product

Catalog No. SQCST27-100B

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Quality Control Standard 27
93.0,3.14,0.05,0.03,0.02,0.02
Liquid
Clear light colored
7732-18-5,7697-37-2,7784-27-2,7789-02-8,16919-19-0,10043-35-3
97.0,3.27,0.05,0.03,0.02,0.02
Amber poly
100mL

Chemical Identifiers

7732-18-5,7697-37-2,7784-27-2,7789-02-8,16919-19-0,10043-35-3
97.0,3.27,0.05,0.03,0.02,0.02
Quality Control Standard 27
93.0,3.14,0.05,0.03,0.02,0.02
Liquid

Safety and Handling

ShelfLife : 12 months