Please login to your online account to display your discounted pricing

Quality Control Standard 27, Ricca Chemical

$2,344.73 - $2,344.73

Chemical Identifiers

Chemical Name or Material Quality Control Standard 27
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 3.14
CAS Max % 3.27
CAS 7784-27-2
CAS Min % 0.05
CAS Max % 0.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SQCSTD27500

 
ricca chemical company
RISQCSTD27500B
Amber poly 500mL Each for $2,344.73

SQCST27-100B

 
ricca chemical company
RISQCSTD27100B
Amber poly 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear light colored
Physical Form Liquid
Quantity 500mL
Chemical Name or Material Quality Control Standard 27
Shelf Life 12 months
CAS Max % 0.05
CAS 7789-02-8
CAS Min % 0.03
CAS Max % 0.03
CAS 16919-19-0
CAS Min % 0.02
CAS Max % 0.02
CAS 10043-35-3
CAS Min % 0.02
CAS Max % 0.02
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
Packaging Amber poly
CAS Min % 3.14
CAS Max % 3.27
CAS 7784-27-2
CAS Min % 0.05
Safety and Handling

12 months