Please login to your online account to display your discounted pricing

Alfa Aesar™ QuadraSil™ SA, metal scavenger

Chemical Identifiers

Chemical Name or Material QuadraSil™ SA
Name Note Metal scavenger
UN Number UN3077
Synonym QuadraSil™ Sulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4726406

  SDS
alfa aesar™
4726406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4726414

  SDS
alfa aesar™
4726414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4726422

  SDS
alfa aesar™
4726422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material QuadraSil™ SA
Name Note Metal scavenger
Physical Form Beads
UN Number UN3077
Synonym QuadraSil™ Sulfonic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.