Alfa Aesar™ QuadraSil™ MP, 20-100 micron, metal scavenger
Metal scavenger: Pd (with or without phosphines), Rh, Cu, Ru, Pt, Pb, Ag, Hg
Manufacturer:  Alfa Aesar™ 4625914
 View more versions of this product
Catalog No. AA4625914
Specifications
Specifications
QuadraSil™ MP metal scavenger
MFCD09753898
Powder or Beads
White to Cream
1225327-73-0
QuadraSil™ Mercaptopropyl
20 to 100 Micron
25g
Chemical Identifiers
1225327-73-0
MFCD09753898
QuadraSil™ Mercaptopropyl
Safety and Handling
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
SDS Documents