Alfa Aesar™ QuadraPure™ TU, 400-600 micron, metal scavenger
Metal Scavenger: Pd, Pt, Ru, Rh, Au, Ag, Cu, Hg, Pb, Cd, Ni, Co, Fe, V, Zn
Manufacturer:  Alfa Aesar™ 4589806
 View more versions of this product
Catalog No. AA4589806
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
QuadraPure™ TU metal scavenger
MFCD07785601
Beads
White to Brown
147754-40-3
QuadraPure™ Thiourea
400 to 600 Micron
5g
Chemical Identifiers
147754-40-3
MFCD07785601
QuadraPure™ Thiourea
SDS Documents