Please login to your online account to display your discounted pricing

Alfa Aesar™ QuadraPure™ MPA, 100-400 micron, metal scavenger

Metal Scavenger: Pd, Ru, Rh, Hg, Au, Ag, Cu, Ni, Sn, Pb, Pt, Cd

$160.08 - $160.08

Chemical Identifiers

Chemical Name or Material QuadraPure™ MPA metal scavenger
MDL Number MFCD07785600
UN Number UN3175
Synonym QuadraPure™ Mercaptophenylaminobut-2-enoate ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4619806

 
alfa aesar™
4619806
5g Each for $160.08
Description & Specifications

Specifications

Synonym QuadraPure™ Mercaptophenylaminobut-2-enoate ester
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Chemical Name or Material QuadraPure™ MPA metal scavenger
TSCA TSCA
Physical Form Beads
MDL Number MFCD07785600
UN Number UN3175
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Particle Size 100 to 400 Micron
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: SOLIDS CONTAINING FLAMMABLE LIQUID, N.O.S.
Color White to yellow
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.