Please login to your online account to display your discounted pricing
Qsonica Cup Horns
Ideal for sterile or pathogenic sample processing
$3,250.86 - $3,250.86
Specifications
For Use With (Equipment) Q700 Sonicator™ without probe
For Use With (Application) Ideal for sterile or pathogenic sample processing
View More Specs
Products
Catalog Number Mfr. No. Holds Type Price Quantity    
15-338-316
qsonica
431C2
20 tubes Cup Horn, Includes sample rack for part#440 Each for $3,250.86
Description & Specifications
Specifications
For Use With (Equipment) Q700 Sonicator™ without probe
For Use With (Application) Ideal for sterile or pathogenic sample processing
  • Multiple samples can be processed
  • Offers indirect sonication and function as high intensity ultrasonic water baths
  • Avoids cross contamination
  • Horn is mounted in acrylic cup
  • Available in two sizes: 3 in. or 5.5 in.
  • Replacement microtube racks sold separately (Cat. Nos. 15-338-404 and -405)