Please login to your online account to display your discounted pricing

QS 11, Tocris

ARFGAP1 inhibitor; modulates Wnt/β-catenin signaling

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material QS 11
Assay Percent Range >98%
Molecular Formula C36H33N5O2
Formula Weight 567.68
Synonym (2S)-2-[2-(Indan-5-yloxy)-9-(1, 1'-biphenyl-4-yl)methyl)-9H-purin-6-ylamino]-3-phenyl-propan-1-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

332410

 
r & d systems
332410
10mg Each for $244.81

332450

 
r & d systems
332450
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 567.68
Quantity 10mg
Chemical Name or Material QS 11
Assay Percent Range >98%
Molecular Formula C36H33N5O2
CAS 944328-88-5
Synonym (2S)-2-[2-(Indan-5-yloxy)-9-(1, 1'-biphenyl-4-yl)methyl)-9H-purin-6-ylamino]-3-phenyl-propan-1-ol
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C