Please login to your online account to display your discounted pricing

QRP™ Qualatrile™ Disposable Nitrile Gloves

  Protects from a wide range of chemicals for splash or intermittent contact

$21.76 - $21.76

Specifications

Color Blue
Length (English) 9 in.
Length (Metric) 22.86cm
Material Latex, natural rubber
Material Vinyl
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-048-740D

 
qrp™
BQF09L
Large Pack of 100 Each for $21.76
Case of 10 Pack for $208.40
Description & Specifications

Specifications

Color Blue
Length (English) 9 in.
Length (Metric) 22.86cm
Material Latex, natural rubber
Material Vinyl
Exterior Finish Textured
Powdered Powder-free
Thickness (English) 5 mil

Nitrile is tough and durable with reduced risk of allergy compared to natural rubber latex.
  • Textured palms and fingers
  • Length: 9 in.
  • Thickness: 5mil
  • Power-free