Please login to your online account to display your discounted pricing

QRP™ Qualatrile™ Blue Nitrile 9 in Powder-Free Gloves

$22.36 - $22.36

Specifications

Certifications ASTM D6319, RoHS
Classification Qualatrile blue Nitrile gloves
Color Blue
Length (English) 9 in.
Length (Metric) 22.86cm
View More Specs
Products
Catalog Number Mfr. No. Thickness Size Price Quantity    

19-048-740A

 
qrp™
BQF09XS
4mil X-Small Pack of 100 Each for $22.36
Case of 10 Pack for $214.10
Description & Specifications

Specifications

Certifications ASTM D6319, RoHS
Classification Qualatrile blue Nitrile gloves
Color Blue
Length (English) 9 in.
Length (Metric) 22.86cm
Material Nitrile, Natural Rubber Latex, Vinyl
Glove Cuff Style Beaded
Hand Ambidextrous
Powdered No