QRP™ Qualatex™ HiPro XC Latex Cleanroom Gloves
The 612HC series rubber lates gloves are 12 in. long and used for Class 100 / ISO 5 cleanroom applications.
Supplier:  QRP™ GP612HCS
 View more versions of this product
Catalog No. 29-635-8397
Alert:
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
  • Premium natural rubber latex gloves for ISO 5 (Class 100) applications
  • Lowest extractibels and particulates
  • Free from plasticizers, silicone and powders
  • Meets or exceeds all applicable requirements of IEST-RP-CC005.2
  • 12 in. Length, Ambidextrous
Specifications
Specifications
IEST-RP-CC005.2
Class 100/ISO 5
No
6 mil
612HCS/PRC
30 cm
Yes
Natural
Small
0.15 mm
12 in.