missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

QRP™ Antistatic Latex Finger Cots

For Class II Static-sensitive devices

Manufacturer:  QRP™ 7JM

 View more versions of this product

Catalog No. 19-081466


Alert:

Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Description

Description

Rolled cot has average surface resistivity of 5 × 1012 ohms/sq. in. Appropriate for Class II (2000-3999V) static-sensitive devices. Can also be used with some higher voltage Class I devices.

  • Made of washed, powder-free latex with proprietary antistatic agent
  • Thickness: 3mil
  • Color: sheer
  • Size: medium

Order Info

Double packed in ESD-safe inner and outer wrap; immobilized in nitrogren-flushed TrayPak™ to prevent charge generation. Packaged 720 cots per pack.

Compliance

Complies with MIL-B-81705, DOD-HDBK-263, and MIL-STD-1686D. ISO Class 5 (formerly Class 100) compatible.

Specifications

Specifications

3mil
Natural Rubber Latex
Antistatic Finger Cot
ASTM, IEST-RP-CC 005.3, MIL-B-81705, DOD-HDBK-263, and MIL-STD-1686D
For use with class II static sensitive devices
Medium
Sheer
SDS
Documents