Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps
Free of odor and taste problems
Manufacturer:  Qorpak™ CAP00456
 View more versions of this product
Catalog No. 36-099-688
Description
Description
  • Made of steel coated with anti-corrosive tinplate
  • Impact resistant
  • Tolerate wide ranges of temperature
  • Feature standard 400 GPI thread finish
  • Used for non-acid, non-alkaline products, organic solvents, chrome cleaners, food bottles and jars, brake fluids and mineral oils
Specifications
Specifications
Pulp/Aluminum Foil
White
1700 Cs.
63-400
Wide mouth bottle
Metal
Bulk