Qorpak™ Valuline Clear Straight Sided Round Bottles with White PP F217 and PTFE Lined Caps
Offers maximum content visibility
$150.49 - $150.49
Specifications
Closure Liner F217 and PTFE
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Clear
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Diameter (English) Closure Size Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Diameter (English) Closure Size Height (English) Price Quantity  
02-992-824 View Documents Qorpak™
239230
32 oz., 960 mL 3.74 in. 89-400 6.69 in. Case of 12 Each for $150.49
Description
Description
  • More economical than other bottles present in market
  • Polypropylene F217 & PTFE lined caps are an economical alternative to Thermoset & Phenolic PTFE lined caps
  • Supplied with attached white Polypropylene F217 & PTFE caps
Specifications
Specifications
F217 and PTFE
Screw
Clear
Polypropylene
White
Glass