Please login to your online account to display your discounted pricing
Qorpak™ ValuLine Amber Boston Round Bottles with White Polypropylene PE Foam/PTFE Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with White Polypropylene PE Foam/PTFE Cap ensures product integrity for long-term storage.
$55.29 - $80.49
Specifications
Closure Liner F217 and PTFE
Closure Material Polypropylene
Closure Type Screw
Color Amber
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Height (Metric) Quantity Price Quantity    
239196
qorpak™
239196
16 oz., 480mL 6.6 in. 168mm 12/Cs. Case of 12 Each for $55.29
239195
qorpak™
239195
8 oz., 240mL 5.43 in. 138mm 24/Cs. Case of 24 Each for $80.49
239191
qorpak™
239191
2 oz., 60mL 3.7 in. 94mm 24/Cs. Case of 24 Each for $57.29
239193
qorpak™
239193
4 oz., 120mL 4.4 in. 112mm 24/Cs. Case of 24 Each for $65.89
239190
qorpak™
239190
32 oz., 960mL 8.2 in. 210mm 12/Cs. Case of 12 Each for $78.23
Description & Specifications
Specifications
Closure Liner F217 and PTFE
Closure Material Polypropylene
Closure Type Screw
Color Amber
Material Glass
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum