Qorpak™ Unlined Round Cans with Triple Tite Lid
Permeation protection against light, oxygen, other gases and essential oils
Manufacturer:  Qorpak™ MET03090
 View more versions of this product
Catalog No. 02-991-229
Description
Description
  • Features resealable multi-friction plug lid for more than one point of contact between lid and can
  • Provides an airtight seal when opened by prying and sealed by pressing
  • May be sterilized by heat
  • Used for paints, varnishes, pastes and adhesives
Specifications
Specifications
3.43 in.
3.875 in.
Triple tite lid and securing clips
Open Top Can
473.1mL
Metal
For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil
314mm
24 Case
16 oz.
Silver
Documents
Accessories