Qorpak™ Safety Coated Amber Jugs — Without Caps
Protects against harmful UV rays
$243.52 - $243.52
Specifications
Closure Type Screw
Closure Size 38-430
Color Amber
Material Plastic Coated Glass
Shape Cylindrical
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
02-992-371 View Documents Qorpak™
GLA08954
2500 mL 6/Cs. Case of 6 Each for $243.52
Description
Description
  • Molded finger grips
  • Slip and impact resistant
  • For handling volatile substances
Specifications
Specifications
Screw
Amber
Cylindrical
38-430
Plastic Coated Glass