Please login to your online account to display your discounted pricing

Qorpak™ PTFE Sealing Septa and Discs


Easy-to-clean, remove and replace

Manufacturer: qorpak™  CAP00542

 View more versions of this product

Catalog No. 02-882-14

  • / Case of 144 Each
 

Description & Specifications

Specifications

Material PTFE
For Use With (Equipment) Bottle closure
Neck Finish 70-400
Packaging Bagged
Thickness (Metric) 0.25mm
Thickness (English) 0.01 in.
Color Natural
No. per Case 144 Cs.