missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ PTFE Sealing Septa and Discs

Easy-to-clean, remove and replace

$1,113.97 - $1,821.61

Specifications

Color Natural
For Use With (Equipment) Bottle closure
View More Specs

Products 2
Catalog Number Mfr. No. Neck Finish No. per Case Price Quantity  
Catalog Number Mfr. No. Neck Finish No. per Case Price Quantity  
02-882-14 View Documents Qorpak™
CAP00542
70-400 144 Cs. Case of 144 Each for $1,113.97
 
02-991-388 View Documents Qorpak™
CAP05188
22 500 Cs. Case of 500 Each for $1,821.61
 
Description

Description

Septa

  • PTFE/Silicone
  • Resists coring
  • Provides great resealability
  • Good for multiple injections
  • PTFE resistant until punctured, once punctured the septa is silicone compatible
Discs
  • Converts ordinary caps into noncontaminating closures

Specifications

Specifications

Natural
Bottle closure