Qorpak™ Polypropylene F422 HDPE Foam Lined Caps
Provides resistance to acids
$1,434.94 - $1,434.94
Specifications
Closure Type Ribbed
Neck Finish 38-439
For Use With (Equipment) Bottle
No. per Case 1100 Cs.
View More Specs
Products 1
Catalog Number Mfr. No. Closure Liner Color Price Quantity  
Catalog Number Mfr. No. Closure Liner Color Price Quantity  
02-991-047 View Documents Qorpak™
CAP00379
F422 HDPE Foam White Case of 1200 Each for $1,434.94
Description
Description
  • Liner is comprised of a 0.38 in. LDPE core sandwiched between identical layers of 0.002 to 0.003 in. virgin HDPE
Specifications
Specifications
Ribbed
38-439
Bottle
1100 Cs.