Please login to your online account to display your discounted pricing
Qorpak™ Natural HDPE Straight Sided Round Bottles — With White Polypropylene SturdeeSeal™ PE Foam Lined Caps
Extra wide opening and jar style allow for easy insertion and recovery of contents
$84.52 - $572.64
Specifications
Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Material Polypropylene
Closure Color White
Closure Size 89-400
Closure Type Screw
View More Specs
Products
Catalog Number Mfr. No. Capacity Packaging Type Quantity Price Quantity    
02-991-318
qorpak™
PLC03421
16 oz., 480mL Conventional Pack 24/Cs. Case of 24 Each for $139.42
02-991-409
qorpak™
PLC03530
32.46 oz., 960mL Bulk Pack 84/Cs. Case of 84 Each for $572.64
02-991-868
qorpak™
PLC03436
32.46 oz., 960mL Conventional Pack 12/Cs. Case of 12 Each for $84.52
Description & Specifications
Specifications
Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Material Polypropylene
Closure Color White
Closure Size 89-400
Closure Type Screw
Color Natural
Material HDPE
Neck Finish 89-400