Please login to your online account to display your discounted pricing
Qorpak™ Natural HDPE Commercial Cylinder Bottles With White PP SturdeeSeal™ PE Foam Caps
Provide excellent moisture barrier
$411.24 - $1,244.64
Specifications
Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Natural
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity    
02-992-917
qorpak™
PLC03389
2 oz., 60mL 24-410 Bulk Pack 900/Cs. Case of 900 Each for $1,244.64
02-993-009
qorpak™
PLC03432
16 oz., 480mL 28-400 Bulk Pack 153/Cs. Case of 153 Each for $411.24
02-993-251
qorpak™
PLC04831
8 oz., 240mL 24-410 Bulk Pack 336/Cs. Case for $656.82
Description & Specifications
Specifications
Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Natural
Material HDPE
  • For handling liquid samples in lab
  • Shoulder style allows to pour small quantities without spillage
  • Available in two different sizes of screw caps: 24mm-410 and 28mm-400
High-speed filling, pharmaceutic specifications and rough handling of interstate shipping