Please login to your online account to display your discounted pricing

Qorpak™ LDPE Cubitainers with White PP SturdeeSeal™ PE Foam Lined Caps

Non-glug construction facilitates flow of contents om continuous uninterrupted streams

$353.41 - $3,007.12

Specifications

Color Natural
Autoclavable No
Material LDPE
Product Type Cubitainer
For Use With (Application) For storage and transport of liquid
View More Specs
Products
Catalog Number Mfr. No. Capacity No. per Case Price Quantity    

02-991-315

 
qorpak™
PLA03242
128 oz., 3.7L 10/Cs. Case of 10 Each for $353.41

02-991-421

 
qorpak™
PLA03243
640 oz., 19L 48/Cs. Case of 48 Each for $3,007.12
Description & Specifications

Specifications

Color Natural
Autoclavable No
Material LDPE
Product Type Cubitainer
For Use With (Application) For storage and transport of liquid
Shape Cube

  • Designed to collapse as product is dispensed
  • Reversible well design permits opening to be raised or depressed for pouring and storage
  • Can withstand rigors of hazardous chemical packaging
  • 128 and 640 oz. (3.7 and 19L) containers include heavy-duty corrugated outer carton