Qorpak™ Kaptclean™ Clear Standard Wide Mouth Bottles, Cleaned for Semi-Volatiles
Wide mouth facilitates easy access to bottle contents
$180.14 - $241.43
Specifications
Closure Liner PTFE
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Clear
View More Specs
Includes: Certificate of Analysis with stock and lot numbers
Products 2
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
02-992-887 View Documents Qorpak™
GLC01737
120 mL 24/Cs. Case of 24 Each for $241.43
02-992-830 View Documents Qorpak™
GLC01825
960 mL 12/Cs. Case of 12 Each for $180.14
Description
Description
  • Made of glass
  • Available with caps made of black polypropylene and PTFE
  • Available in volumes ranging from 4 to 32 oz. (120 to 960mL)
Certifications
Precleaned to meet U.S. EPA Specifications and Guidelines for Contaminant-Free Sample Containers
Specifications
Specifications
PTFE
Screw
Clear
Phenolic
Black
Glass