Qorpak™ Amber Wide Mouth Packer Bottles with Polypropylene Cap and PTFE Disc
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Glass Jug with Polypropylene Cap and PTFE Disc are used in chemical and environmental applications.
$145.94 - $145.94
Specifications
Closure Liner PTFE
Closure Material Polypropylene
Closure Type Screw
Closure Color Black
Color Amber
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity  
02-991-824 View Documents Qorpak™
GLC02155
32 oz., 950 mL 53-400 Qorpak Carton 12/Cs. Case of 12 Each for $145.94
Description
Description
  • Feature a wide opening for making them the ideal general purpose bottle for solids
  • Available with caps made of black polypropylene and friction fit 0.015 in. (0.38mm) PTFE disc
  • Both cap and liner can be washed, heated or autoclaved
  • Available in volumes ranging from 1 to 32 oz. (30 to 960mL)
Specifications
Specifications
PTFE
Screw
Amber
Polypropylene
Black
Glass