Qorpak™ HDPE Pail Lids
For pails of 1, 2, 3.5, 5 and 6 gal. volumes
Manufacturer:  Qorpak™ 240126
 View more versions of this product
Catalog No. 02-991-162
Description
Description
  • Available in plain, Senior Rieke spout and EZ Tear styles
Rieke spout and Senior Rieke spout lids:
  • Facilitate dispensing of liquid contents
EZ Tear lids:
  • Feature pull tabs that secure contents during shipping and transport
Specifications
Specifications
White
100/Cs.
HDPE
Plain Lid
Documents