missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ HDPE Open Head Pails

Suited for general or warehouse usage

Manufacturer:  Qorpak™ 240131

 View more versions of this product

Catalog No. 02-991-051


 

Specifications Description & Specifications

Specifications

5 gal., 18.9L
10.5 in.
26.67cm
Metal
37.16cm
10/Cs.
Open Head Pail
18.9L
White
11.88 in.
30.17cm
14.63 in.
Embossed safety warning label, rounded hand grip and metal bail
HDPE
5 gal.