Please login to your online account to display your discounted pricing

Qorpak™ Green Thermoset F217 and PTFE Lined Caps


Offer wide range of chemical compatibility and high temperature tolerance

Manufacturer: qorpak™  CAP00560

 View more versions of this product

Catalog No. 02-993-422

  • / Case of 500 Each
 

Description & Specifications

Specifications

Neck Finish 33-400
Closure Type Smooth
For Use With (Equipment) Wide mouth bottle
Packaging Bagged
Color Green
No. per Pack 100/Pk.
Closure Liner F217 and PTFE
No. per Case 500 Cs.