Please login to your online account to display your discounted pricing

Qorpak™ Green Thermoset F217 and PTFE Lined Caps


Offer wide range of chemical compatibility and high temperature tolerance

Manufacturer: qorpak™  CAP00553

 View more versions of this product

Catalog No. 02-991-195

  • / Case
 

Description & Specifications

Specifications

For Use With (Equipment) Wide mouth bottle
Neck Finish 22-400
Closure Type Smooth
Closure Liner F217 and PTFE
Packaging Bulk
Color Green
No. per Case 4600 Cs.