Please login to your online account to display your discounted pricing

Qorpak™ Green Thermoset F217 and PTFE Lined Caps


Offer wide range of chemical compatibility and high temperature tolerance

Manufacturer: qorpak™  CAP00555

 View more versions of this product

Catalog No. 02-991-148

  • / Case
 

Description & Specifications

Specifications

Closure Liner F217 and PTFE
Closure Type Smooth
Packaging Bulk
Color Green
For Use With Wide mouth bottle
Neck Finish 24-400
No. per Case 4000 Cs.