Please login to your online account to display your discounted pricing

Qorpak™ Green Thermoset F217 and PTFE Lined Caps


Offer wide range of chemical compatibility and high temperature tolerance

Manufacturer: qorpak™  CAP00551

 View more versions of this product

Catalog No. 02-991-147

  • / Case
 

Description & Specifications

Specifications

Neck Finish 20-400
For Use With (Equipment) Wide mouth bottle
Closure Type Smooth
Color Green
No. per Case 4800 Cs.
Closure Liner F217 and PTFE
Packaging Bulk