Qorpak™ Graduated Medium Round Bottle Beakers™ without Caps
For mixing, measuring and storage
Supplier: Qorpak™ GLA00843
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
83 mm | |
42 mm | |
38-400 mm | |
60 mL | |
Clear | |
Glass |
Screw Cap | |
3.27 in. | |
2 oz., 60 mL | |
2 oz. | |
1.65 in. | |
48/Cs. |