Qorpak™ Graduated Medium Round Bottle Beakers™ without Caps
For mixing, measuring and storage
Manufacturer:  Qorpak™ GLA00844
 View more versions of this product
Catalog No. 02991796
Description
Description
  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents
Specifications
Specifications
4 oz., 120mL
4 oz.
2.01 in.
Glass
48-400
102mm
120mL
Clear
4.02 in.
144/Cs.
51mm
Screw
Documents