Qorpak™ Graduated Medium Round Bottle Beakers™ without Caps
For mixing, measuring and storage
Manufacturer:  Qorpak™ GLA00848
 View more versions of this product
Catalog No. 02991732
Description
Description
  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents
Specifications
Specifications
16 oz., 480mL
16 oz.
2.99 in.
Glass
70-400
145mm
480mL
Clear
5.71 in.
48/Cs.
76mm
Screw
Documents