missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Solid PE Caps

For mixing, measuring and storage

Manufacturer:  Qorpak™ GLC01462

 View more versions of this product

Catalog No. 02992139


 

Description

Description

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents

Specifications

Specifications

1 oz., 30mL
1 oz.
Polyethylene
1.34 in.
Glass
48/Cs.
33-400
68mm
30mL
Phenolic
Clear
2.68 in.
Conventional Pack
Black
33mm
Screw
Documents