missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Rubber Lined Caps

For mixing, measuring and storage

Manufacturer:  Qorpak™ GLC01478

 View more versions of this product

Catalog No. 02992453


 

Description

Description

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents

Specifications

Specifications

2 oz., 60mL
2 oz.
Rubber
1.65 in.
Glass
288/Cs.
38-400
83mm
60mL
Phenolic
Clear
3.27 in.
Bulk Pack
Black
42mm
Screw
Documents